Gene Transcripts

Gene Symbol
SRL
Entrez Gene ID
6345
Gene Name
sarcalumenin
Gene Aliases
Chromosome Location
Chr. 16: 4189374 - 4242253 on Build GRCh38
UniGene ID
Hs.10041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098814.1 NP_001092284.1 6 673
NM_001323667.1 NP_001310596.1 7 803
NM_001323668.1 NP_001310597.1 7 2051
XM_017023527.1 XP_016879016.1 7 1998
XM_017023528.1 XP_016879017.1 6 704
XM_017023529.1 XP_016879018.1 6 719
GenBank mRNA AK056588.1 - 6 673
AK302601.1 - 6 709
AL831973.2 - 6 645
AL832361.1 - 7 2045
BX647476.1 - 7 2045

View Full Product Details