Gene Transcripts

Gene Symbol
FRMPD3
Entrez Gene ID
84443
Gene Name
FERM and PDZ domain containing 3
Gene Aliases
Chromosome Location
Chr. X: 107522450 - 107605264 on Build GRCh38
UniGene ID
Hs.496546
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032428.1 NP_115804.1 9 799
XM_006724709.2 XP_006724772.1 3 274
XM_011531056.2 XP_011529358.1 7 775
XM_011531057.2 XP_011529359.1 7 701
XM_011531058.2 XP_011529360.1 6 613
XM_017029901.1 XP_016885390.1 7 1020
XM_017029902.1 XP_016885391.1 7 1020

View Full Product Details