Gene Transcripts

Gene Symbol
KSR1
Entrez Gene ID
8844
Gene Name
kinase suppressor of ras 1
Gene Aliases
KSR, RSU2
Chromosome Location
Chr. 17: 27456441 - 27626438 on Build GRCh38
UniGene ID
Hs.133534
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014238.1 NP_055053.1 22 4424
XM_006722154.3 XP_006722217.1 19 3083
XM_011525429.2 XP_011523731.1 21 3191
XM_011525430.2 XP_011523732.1 21 4578
XM_017025267.1 XP_016880756.1 20 3149
XM_017025268.1 XP_016880757.1 20 3125
XM_017025270.1 XP_016880759.1 20 2926
XM_017025271.1 XP_016880760.1 22 3898
XM_017025272.1 XP_016880761.1 22 3881
XM_017025273.1 XP_016880762.1 21 3704
XM_017025274.1 XP_016880763.1 21 2907
XM_017025275.1 XP_016880764.1 21 2894
XM_017025276.1 XP_016880765.1 22 3846
XM_017025277.1 XP_016880766.1 17 2589
XM_017025278.1 XP_016880767.1 17 2599
GenBank mRNA AK124344.1 - 1 91

View Full Product Details