Gene Transcripts

Gene Symbol
KRTAP6-3
Entrez Gene ID
337968
Gene Name
keratin associated protein 6-3
Gene Aliases
KAP6.3
Chromosome Location
Chr. 21: 30592440 - 30593055 on Build GRCh38
UniGene ID
Hs.553691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181605.3 NP_853636.3 1 563
GenBank mRNA AB096951.1 - 1 563

View Full Product Details