Gene Transcripts

Gene Symbol
FAM21A
Entrez Gene ID
387680
Gene Name
family with sequence similarity 21 member A
Gene Aliases
FAM21B, bA56A21.1, bA98I6.1
Chromosome Location
Chr. 10: 50067888 - 50133509 on Build GRCh38
UniGene ID
Hs.365286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005751.2 NP_001005751.1 27 2963
XM_005269806.3 XP_005269863.2 26 2839
XM_005269807.3 XP_005269864.2 26 2824
XM_005269810.3 XP_005269867.2 9 1056
XM_006717831.3 XP_006717894.2 26 2758
XM_017016227.1 XP_016871716.1 26 2788
XM_017016230.1 XP_016871719.1 26 2776
XM_017016233.1 XP_016871722.1 24 2486
XM_017016234.1 XP_016871723.1 26 2846
XM_017016235.1 XP_016871724.1 18 1862
GenBank mRNA AK001686.1 - 24 2615
AK304116.1 - 27 2899
BC082258.1 - 27 2890 6278433
CN314884.1 - 4 288

Gene Symbol
FAM21C
Entrez Gene ID
253725
Gene Name
family with sequence similarity 21 member C
Gene Aliases
FAM21A, VPEF
Chromosome Location
Chr. 10: 45727170 - 45792964 on Build GRCh38
UniGene ID
Hs.365286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011539566.1 XP_011537868.1 27 2930
XM_011539567.1 XP_011537869.1 26 2858
XM_011539568.1 XP_011537870.1 26 2843
XM_011539569.1 XP_011537871.1 26 2777
XM_011539570.2 XP_011537872.1 26 2819
XM_011539573.2 XP_011537875.1 9 1056
XM_017016014.1 XP_016871503.1 26 2807
XM_017016017.1 XP_016871506.1 25 2771
XM_017016019.1 XP_016871508.1 25 2705
XM_017016021.1 XP_016871510.1 27 2858
XM_017016024.1 XP_016871513.1 15 1672
XM_017016025.1 XP_016871514.1 16 1743
GenBank mRNA AK293929.1 - 22 2414
AL050279.1 - 1 276

View Full Product Details