Gene Transcripts

Gene Symbol
SPDYC
Entrez Gene ID
387778
Gene Name
speedy/RINGO cell cycle regulator family member C
Gene Aliases
RINGOC, Ringo2
Chromosome Location
Chr. 11: 65170236 - 65173666 on Build GRCh38
UniGene ID
Hs.450642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008778.1 NP_001008778.1 2, 3 199
XM_017017705.1 XP_016873194.1 1, 2 593
XM_017017706.1 XP_016873195.1 1, 2 593
XM_017017707.1 XP_016873196.1 1, 2 593
XM_017017708.1 XP_016873197.1 1, 2 593
XM_017017709.1 XP_016873198.1 1, 2 593
XM_017017710.1 XP_016873199.1 1, 2 593
GenBank mRNA AY820305.1 - 2, 3 199
BC137244.1 - 2, 3 281 9021241
BC137246.1 - 2, 3 281 9021243
BC144548.1 - 2, 3 281 9053075

View Full Product Details