Gene Transcripts

Gene Symbol
PAGE2B
Entrez Gene ID
389860
Gene Name
PAGE family member 2B
Gene Aliases
CT16.5, GAGEE3, PAGE-2B
Chromosome Location
Chr. X: 55027927 - 55078903 on Build GRCh38
UniGene ID
Hs.293317
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015038.2 NP_001015038.1 2 84
XM_011530785.2 XP_011529087.1 3 500
XM_011530786.2 XP_011529088.1 2 167
XM_011530787.2 XP_011529089.1 2 147
XM_017029513.1 XP_016885002.1 2 104
GenBank mRNA BC130359.1 - 2 43
BM782405.1 - 2 69

View Full Product Details