Gene Transcripts

Gene Symbol
RNASE13
Entrez Gene ID
440163
Gene Name
ribonuclease A family member 13 (inactive)
Gene Aliases
HEL-S-86p, RAL1
Chromosome Location
Chr. 14: 21032820 - 21034785 on Build GRCh38
UniGene ID
Hs.666729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012264.3 NP_001012264.1 2 268
GenBank mRNA AY665808.1 - 1 130
BC044831.1 - 2 268 5166630
FJ237363.1 - 2 268

View Full Product Details