Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_002824.3 | 32 | 4892 | |||
GenBank mRNA | AB002391.2 | - | 29 | 4579 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
32 | 4898 | ||
AF041081.1 | - | LIT2191 HERC2P3 |
28 | 4855 | ||
AK095014.1 | - | 8 | 2020 | |||
CR749409.1 | - | 21 | 3334 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_036443.1 | 10 | 1517 | ||
GenBank mRNA | AK293512.1 | - | 10 | 1568 | |
BC047911.1 | - | 10 | 1517 |