Gene Transcripts

Gene Symbol
FBXO43
Entrez Gene ID
286151
Gene Name
F-box protein 43
Gene Aliases
EMI2, ERP1, FBX43
Chromosome Location
Chr. 8: 100133360 - 100150569 on Build GRCh38
UniGene ID
Hs.339577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029860.3 NP_001025031.2 5 2863
NR_036491.1 5 2907
XM_011516987.2 XP_011515289.1 6 2938
XM_011516989.2 XP_011515291.1 6 2982
XM_011516990.1 XP_011515292.1 6 2208
GenBank mRNA BC090892.1 - 5 2836
BG391951.1 - 1 112

View Full Product Details