Gene Transcripts

Gene Symbol
UBQLN4
Entrez Gene ID
56893
Gene Name
ubiquilin 4
Gene Aliases
A1U, A1Up, C1orf6, CIP75, UBIN
Chromosome Location
Chr. 1: 156035294 - 156053825 on Build GRCh38
UniGene ID
Hs.283739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304342.1 NP_001291271.1 1 201
NM_020131.4 NP_064516.2 1 201
XM_005245348.3 XP_005245405.1 1 106
GenBank mRNA AB209566.1 - 1 74
AF188240.1 - 1 355
AK098368.1 - 1 89
AK302895.1 - 1 90
AK314413.1 - 1 89
BC018403.2 - 1 89 4214386
BC063841.1 - 1 101 6183942
DC334851.1 - 1 184
HY061664.1 - 1 205

View Full Product Details