Gene Transcripts

Gene Symbol
Vom1r54
Entrez Gene ID
365190
Gene Name
vomeronasal 1 receptor 54
Gene Aliases
V1rl1, V1rm4
Chromosome Location
Chr. 1: 66388887 - 66389811 on Build Rnor_6.0
UniGene ID
Rn.129413
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008963.1 NP_001008963.1 886
GenBank mRNA AY510374.1 - 886

View Full Product Details