Gene Transcripts

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
Chromosome Location
Chr. 10: 50184861 - 50251564 on Build GRCh38
UniGene ID
Hs.512645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143974.1 NP_001137446.1 2 345
NM_019893.2 NP_063946.2 2 345
XM_011539970.2 XP_011538272.1 3 541
XM_011539971.2 XP_011538273.1 3 545
XM_011539972.2 XP_011538274.1 3 542
XM_011539973.2 XP_011538275.1 3 551
XM_017016435.1 XP_016871924.1 3 545
GenBank mRNA AF250847.1 - 2 288
AF449759.2 - 2 345
BC107105.2 - 2 288 40018929

View Full Product Details