Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025687.1 | NP_001020858.1 | 5 | 1112 | |
XM_006233078.2 | XP_006233140.1 | 4 | 1194 | ||
XM_006233079.1 | XP_006233141.1 | 5 | 1107 | ||
XM_006233080.2 | XP_006233142.1 | 3 | 1013 | ||
GenBank mRNA | BC098939.1 | - | 5 | 1112 |