Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025425.1 | NP_001020596.1 | 3, 4 | 341 | |
XM_006248768.2 | XP_006248830.1 | 4, 5 | 526 | ||
XM_006248769.1 | XP_006248831.1 | 4, 5 | 531 | ||
XM_008768985.1 | XP_008767207.1 | 3, 4 | 482 | ||
GenBank mRNA | AY879265.1 | - | 3, 4 | 341 | |
GU264001.1 | - | 4, 5 | 389 |