Gene Transcripts

Gene Symbol
Mks1
Entrez Gene ID
287612
Gene Name
Meckel syndrome, type 1
Gene Aliases
Chromosome Location
Chr. 10: 75149782 - 75160825 on Build Rnor_6.0
UniGene ID
Rn.198890
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034917.2 NP_001030089.2 13, 14 1169
GenBank mRNA BC099806.1 - 13, 14 1167

View Full Product Details