Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128427.2 | NP_001121899.1 | 3, 4 | 601 | |
NM_001128428.2 | NP_001121900.1 | 3, 4 | 601 | ||
NM_020116.4 | NP_064501.2 | 3, 4 | 604 | ||
XM_011532126.1 | XP_011530428.1 | 3, 4 | 570 | ||
GenBank mRNA | AB033089.1 | - | 3, 4 | 531 | |
AK315152.1 | - | 3, 4 | 572 | ||
BC036502.2 | - | 3, 4 | 605 | 5265170 | |
BC073978.1 | - | 3, 4 | 304 | ||
DA243182.1 | - | 3, 4 | 587 |