Gene Transcripts

Gene Symbol
Kdm8
Entrez Gene ID
308976
Gene Name
lysine (K)-specific demethylase 8
Gene Aliases
Jmjd5, RGD1304823
Chromosome Location
Chr. 1: 196838457 - 196854192 on Build Rnor_6.0
UniGene ID
Rn.22093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037196.1 NP_001032273.1 8 1440
XM_006230258.2 XP_006230320.1 8 2127
XM_006230259.2 XP_006230321.1 7 2211
GenBank mRNA BC100627.1 - 8 1440

View Full Product Details