Gene Transcripts

Gene Symbol
Adamts10
Entrez Gene ID
314655
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif, 10
Gene Aliases
Chromosome Location
Chr. 7: 18409068 - 18439012 on Build Rnor_6.0
UniGene ID
Rn.135600
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271445.1 NP_001258374.1 3 378
GenBank mRNA BC101863.1 - 3 347

View Full Product Details