Gene Transcripts

Gene Symbol
Bex4
Entrez Gene ID
501624
Gene Name
brain expressed, X-linked 4
Gene Aliases
Chromosome Location
Chr. X|NW_007906392: 30532 - 31123 on Build Rnor_6.0
UniGene ID
Rn.9287
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037554.1 NP_001032643.1 1 637
GenBank mRNA AY833557.1 - 1 637
BC128698.1 - 1 728 8363772

View Full Product Details