Gene Transcripts

Gene Symbol
Defa11
Entrez Gene ID
613225
Gene Name
defensin alpha 11
Gene Aliases
Chromosome Location
Chr. 16: 75517421 - 75519605 on Build Rnor_6.0
UniGene ID
Rn.215839
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033078.1 NP_001028250.1 2 183
XM_006253369.1 XP_006253431.1 3 281
GenBank mRNA AY623755.1 - 2 183

View Full Product Details