Gene Transcripts

Gene Symbol
Scgb2b26
Entrez Gene ID
110187
Gene Name
secretoglobin, family 2B, member 26
Gene Aliases
Abpbg26, Abpbg4, Abpg, C2d, Scgb2b4
Chromosome Location
Chr. 7: 33942997 - 33944985 on Build GRCm38
UniGene ID
Mm.155750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178308.2 NP_840093.1 3 289
GenBank mRNA AY293281.1 - 3 285
AY293282.1 - 2 199
BC035222.1 - 3 275 4192901
BC107292.1 - 3 277 40053055
BC107293.1 - 3 277 40053056
BC111446.1 - 3 289 40053210
BC134362.1 - 3 295 40134659
BC148302.2 - 3 295 40134662

View Full Product Details