Gene Transcripts

Gene Symbol
Orm3
Entrez Gene ID
18407
Gene Name
orosomucoid 3
Gene Aliases
Agp-3, Orm-3
Chromosome Location
Chr. 4: 63355279 - 63359511 on Build GRCm38
UniGene ID
Mm.381399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013623.2 NP_038651.2 1 106
GenBank mRNA BC141235.1 - 1 104
BC145559.1 - 1 104 9054123
BC145560.1 - 1 104
BC171932.1 - 1 104

View Full Product Details