Gene Transcripts

Gene Symbol
Ankrd12
Entrez Gene ID
106585
Gene Name
ankyrin repeat domain 12
Gene Aliases
2900001A12Rik, AI447928, ANCO-2, AV347965, GAC-1, mKIAA0874
Chromosome Location
Chr. 17: 65965703 - 66077046 on Build GRCm38
UniGene ID
Mm.34706
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025572.1 NP_001020743.1 9 1788
GenBank mRNA AK046455.1 - 1 112
AK134530.1 - 4 967
CF553222.1 - 1 210

View Full Product Details