Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033310.3 | NP_001028482.2 | 2 | 428 | |
NM_001163456.1 | NP_001156928.1 | 2 | 428 | ||
GenBank mRNA | AK156053.1 | - | 2 | 397 | |
BC038311.1 | - | 2 | 198 | ||
BC138972.1 | - | 2 | 406 | 8861994 | |
BC145479.1 | - | 2 | 406 | 9054042 | |
BC145486.1 | - | 2 | 410 | ||
BC145728.1 | - | 2 | 406 | 40131031 | |
CJ085333.1 | - | 2 | 376 | ||
CX225681.1 | - | 2 | 355 | ||
DN174415.1 | - | 2 | 403 | ||
DV661727.1 | - | 1 | 39 |