Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033463.3 | NP_001028635.2 | 3 | 796 | |
GenBank mRNA | AK146408.1 | - | 2 | 545 | |
AK156197.1 | - | 3 | 797 | ||
AK220217.1 | - | 2 | 727 | ||
BC141065.1 | - | 3 | 731 | 9055677 | |
BC145318.1 | - | 3 | 731 | 9053873 |