Gene Transcripts

Gene Symbol
PDXP
Entrez Gene ID
57026
Gene Name
pyridoxal phosphatase
Gene Aliases
CIN, PLP, dJ37E16.5
Chromosome Location
Chr. 22: 37658730 - 37666932 on Build GRCh38
UniGene ID
Hs.632762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020315.4 NP_064711.1 2 1406
GenBank mRNA AK057265.1 - 2 972
BC000320.2 - 2 1405 2821743
BC009756.2 - 2 995
BC018844.2 - 2 1374
BC064922.1 - 2 1406 6141538

View Full Product Details