Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 70 | |
GenBank mRNA | BC074991.2 | - | 1 | 68 | |
BC074992.2 | - | 1 | 68 | 30915307 | |
BC113994.1 | - | 1 | 70 | 40084821 | |
BC114000.1 | - | 1 | 70 | 40084820 |