Gene Transcripts

Gene Symbol
KLRC2
Entrez Gene ID
3822
Gene Name
killer cell lectin like receptor C2
Gene Aliases
CD159c, NKG2-C, NKG2C
Chromosome Location
Chr. 12: 10430599 - 10435993 on Build GRCh38
UniGene ID
Hs.591157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002260.3 NP_002251.2 6 717
GenBank mRNA BC106069.1 - 6 669

View Full Product Details