Gene Transcripts

Gene Symbol
PDSS1
Entrez Gene ID
23590
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
Gene Aliases
COQ1, COQ10D2, DPS, SPS, TPRT, TPT, TPT 1, hDPS1
Chromosome Location
Chr. 10: 26697052 - 26746798 on Build GRCh38
UniGene ID
Hs.558468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321978.1 NP_001308907.1 3 482
NM_001321979.1 NP_001308908.1 3 482
NM_014317.4 NP_055132.2 3 482
XM_017016011.1 XP_016871500.1 2 69
GenBank mRNA AB209763.1 - 2 2520
AB210838.1 - 3 240
AK024802.1 - 3 247
AK223414.1 - 3 247
BC049211.1 - 3 192
BC063635.1 - 3 210 4397753

View Full Product Details