Gene Transcripts
- Gene Symbol
- C22orf39
- Entrez Gene ID
- 128977
- Gene Name
- chromosome 22 open reading frame 39
- Gene Aliases
- Chromosome Location
- Chr. 22: 19440886 - 19448232 on Build GRCh38
- UniGene ID
- Hs.659565
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC062599.1 |
- |
3 |
726 |
|
BX648073.1 |
- |
1 |
30 |
|
View Full Product Details