Gene Transcripts

Gene Symbol
C22orf39
Entrez Gene ID
128977
Gene Name
chromosome 22 open reading frame 39
Gene Aliases
Chromosome Location
Chr. 22: 19440886 - 19448232 on Build GRCh38
UniGene ID
Hs.659565
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173793.4 NP_776154.3 3 1075
GenBank mRNA BC062599.1 - 3 540

View Full Product Details