Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010854.1 | NP_001010854.1 | 2, 3 | 391 | |
XM_017021044.1 | XP_016876533.1 | 2, 3 | 355 | ||
XM_017021045.1 | XP_016876534.1 | 2, 3 | 355 | ||
XM_017021047.1 | XP_016876536.1 | 2, 3 | 188 | ||
GenBank mRNA | AL832848.1 | - | 1, 2 | 69 | |
BG489583.1 | - | 1, 2 | 57 | ||
BQ426031.1 | - | 306 | |||
CB988671.1 | - | 418 |