Gene Transcripts

Gene Symbol
SLC5A9
Entrez Gene ID
200010
Gene Name
solute carrier family 5 member 9
Gene Aliases
SGLT4
Chromosome Location
Chr. 1: 48221113 - 48248644 on Build GRCh38
UniGene ID
Hs.37890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011547.2 NP_001011547.2 14 2395
NM_001135181.1 NP_001128653.1 15 2470
XM_011540924.2 XP_011539226.1 14 4076
XM_011540926.2 XP_011539228.1 13 2656
XM_011540927.1 XP_011539229.1 12 2254
XM_011540928.1 XP_011539230.1 14 2274
GenBank mRNA AK131200.1 - 14 2348
BC025411.1 - 5 1195
BX648549.1 - 15 2424

View Full Product Details