Gene Transcripts

Gene Symbol
RT1-CE13
Entrez Gene ID
414790
Gene Name
RT1 class I, locus CE13
Gene Aliases
Chromosome Location
Chr. 20|NW_007906368: 3928 - 4540 on Build Rnor_6.0
UniGene ID
Rn.232675
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008836.1 NP_001008836.1 6 915

View Full Product Details