Gene Transcripts

Gene Symbol
Vom1r79
Entrez Gene ID
494227
Gene Name
vomeronasal 1 receptor 79
Gene Aliases
V1rc35
Chromosome Location
Chr. 4: 87981869 - 87982729 on Build Rnor_6.0
UniGene ID
Rn.129661
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009508.1 NP_001009508.1 1 640
GenBank mRNA AY510284.1 - 1 640

View Full Product Details