Gene Transcripts
- Gene Symbol
- LOC305806
- Entrez Gene ID
- 305806
- Gene Name
- similar to glutaredoxin 1 (thioltransferase); glutaredoxin
- Gene Aliases
- Chromosome Location
- Chr. 15: 5780834 - 5794911 on Build Rnor_6.0
- UniGene ID
- Rn.123447
- Species
- Rattus norvegicus
- Gene Symbol
- LOC290508
- Entrez Gene ID
- 290508
- Gene Name
- similar to RIKEN cDNA 1700001F09
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Rn.159598
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC079266.1 |
- |
|
818 |
|
View Full Product Details