Gene Transcripts

Gene Symbol
Yipf2
Entrez Gene ID
363027
Gene Name
Yip1 domain family, member 2
Gene Aliases
RGD1307512
Chromosome Location
Chr. 8: 22621758 - 22625959 on Build Rnor_6.0
UniGene ID
Rn.233756
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014208.1 NP_001014230.1 3 218
GenBank mRNA BC083587.1 - 3 218

View Full Product Details