Gene Transcripts

Gene Symbol
RSA-14-44
Entrez Gene ID
297173
Gene Name
RSA-14-44 protein
Gene Aliases
Chromosome Location
Chr. 4: 96831880 - 96833252 on Build Rnor_6.0
UniGene ID
Rn.47032
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182670.2 NP_872611.2 1 1331
GenBank mRNA BC083656.1 - 1 1320

View Full Product Details