Gene Transcripts

Gene Symbol
Ctrc
Entrez Gene ID
76701
Gene Name
chymotrypsin C (caldecrin)
Gene Aliases
1810044E12Rik
Chromosome Location
Chr. 4: 141838239 - 141846359 on Build GRCm38
UniGene ID
Mm.489680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033875.2 NP_001029047.1 4, 5 441
GenBank mRNA AK007772.1 - 3, 4 269
BC103715.1 - 4, 5 439 6430373

View Full Product Details