Gene Transcripts

Gene Symbol
Tgoln2
Entrez Gene ID
22135
Gene Name
trans-golgi network protein 2
Gene Aliases
TGN38B, TGN41, TGN46, Ttgn2
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.246563
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009444.1 NP_033470.1 4 1504
GenBank mRNA AK145212.1 - 4 1491
D50032.1 - 4 1504

View Full Product Details