Gene Transcripts

Gene Symbol
Hist1h2ag
Entrez Gene ID
319167
Gene Name
histone cluster 1, H2ag
Gene Aliases
Chromosome Location
Chr. 13: 22042478 - 22042949 on Build GRCm38
UniGene ID
Mm.263165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178186.3 NP_835493.1 1 437
GenBank mRNA BC065803.1 - 1 437

View Full Product Details