Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020997.3 | NP_066277.1 | 4 | 1210 | |
GenBank mRNA | AF081512.1 | - | 4 | 1200 | |
AK129605.1 | - | 4 | 913 | ||
AK222714.1 | - | 4 | 1210 | ||
AY358873.1 | - | 4 | 1197 | ||
BC027883.1 | - | 4 | 1197 | 5221120 |