Gene Transcripts

Gene Symbol
Vmn1r183
Entrez Gene ID
209824
Gene Name
vomeronasal 1 receptor 183
Gene Aliases
Gm476, V1rd15
Chromosome Location
Chr. 7: 24054774 - 24055691 on Build GRCm38
UniGene ID
Mm.377233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203489.1 NP_987074.1 1 20
GenBank mRNA BC130237.1 - 1 24

View Full Product Details