Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011547.2 | NP_001011547.2 | 14 | 2131 | |
NM_001135181.1 | NP_001128653.1 | 15 | 2206 | ||
XM_011540924.2 | XP_011539226.1 | 14 | 3812 | ||
XM_011540926.2 | XP_011539228.1 | 13 | 2392 | ||
XM_011540927.1 | XP_011539229.1 | 12 | 1990 | ||
XM_011540928.1 | XP_011539230.1 | 14 | 2010 | ||
GenBank mRNA | AK131200.1 | - | 14 | 2084 | |
AK311580.1 | - | 4 | 834 | ||
BC025411.1 | - | 5 | 931 | ||
BC104857.1 | - | 14 | 2131 | 8143860 | |
BC104863.1 | - | 14 | 2131 | 8143866 | |
BC143570.1 | - | 15 | 2194 | 9052081 | |
BC143572.1 | - | 15 | 2204 | 9052083 | |
BC143587.1 | - | 13 | 1925 | ||
BX648549.1 | - | 15 | 2160 |