Gene Transcripts

Gene Symbol
ARF5
Entrez Gene ID
381
Gene Name
ADP ribosylation factor 5
Gene Aliases
Chromosome Location
Chr. 7: 127588352 - 127591705 on Build GRCh38
UniGene ID
Hs.653202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001662.3 NP_001653.1 4 421
GenBank mRNA AF493884.1 - 4 274
AK297026.1 - 4 326
AK307297.1 - 3 866
AK308548.1 - 3 1074
AK312107.1 - 4 326
BC003043.1 - 4 311 2822973
BC033104.1 - 4 368 4862014
BT007087.1 - 4 274
CN259625.1 - 4 438
M57567.1 - 4 328

View Full Product Details