Gene Transcripts

Gene Symbol
HOXB7
Entrez Gene ID
3217
Gene Name
homeobox B7
Gene Aliases
HHO.C1, HOX2, HOX2C, Hox-2.3
Chromosome Location
Chr. 17: 48607232 - 48611021 on Build GRCh38
UniGene ID
Hs.436181
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004502.3 NP_004493.3 2 1049
GenBank mRNA AK223249.1 - 2 1047
BQ233410.1 - 1 367
M16937.1 - 2 1045
S49765.1 - 3 591

View Full Product Details