Gene Transcripts

Gene Symbol
NAAA
Entrez Gene ID
27163
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
ASAHL, PLT
Chromosome Location
Chr. 4: 75910655 - 75941013 on Build GRCh38
UniGene ID
Hs.437365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042402.1 NP_001035861.1 4 583
NM_014435.3 NP_055250.2 4 583
XM_005262920.3 XP_005262977.1 4 581
XM_005262921.3 XP_005262978.1 4 581
XM_005262923.2 XP_005262980.1 4 581
XM_005262924.2 XP_005262981.1 4 581
XM_005262925.3 XP_005262982.1 4 581
XM_006714180.2 XP_006714243.1 4 581
XM_011531852.2 XP_011530154.1 4 581
XM_011531854.2 XP_011530156.1 4 581
XM_017008027.1 XP_016863516.1 4 581
XM_017008028.1 XP_016863517.1 4 581
XM_017008029.1 XP_016863518.1 4 581
XM_017008030.1 XP_016863519.1 4 581
GenBank mRNA AB161353.1 - 4 519
AK024677.1 - 1083
AK301132.1 - 3 408
BC006388.2 - 4 563 4122755
BM763257.1 - 4 583
M92449.1 - 3 279

View Full Product Details