Gene Transcripts

Gene Symbol
LRRN1
Entrez Gene ID
57633
Gene Name
leucine rich repeat neuronal 1
Gene Aliases
FIGLER3, NLRR-1
Chromosome Location
Chr. 3: 3799431 - 3849834 on Build GRCh38
UniGene ID
Hs.163244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324188.1 NP_001311117.1 3 2290
NM_001324189.1 NP_001311118.1 3 2213
NM_020873.6 NP_065924.3 2 2008
GenBank mRNA AB040930.1 - 2 4507
AB074161.1 - 1 1241
AK023273.1 - 1 395
AK291717.1 - 2 1592
AY358312.1 - 1 1355
BC034947.2 - 2 1571 4826337

View Full Product Details