Gene Transcripts

Gene Symbol
CRLF2
Entrez Gene ID
64109
Gene Name
cytokine receptor-like factor 2
Gene Aliases
CRL2, CRLF2Y, TSLPR
Chromosome Location
Chr. X: 1190437 - 1212762 on Build GRCh38
UniGene ID
Hs.287729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012288.2 NP_001012288.2 3 362
NM_022148.3 NP_071431.2 4 465
NR_110830.1 4 462
XM_011546181.2 XP_011544483.1 4 500
XM_011546182.2 XP_011544484.1 4 504
GenBank mRNA AB052639.1 - 4 376
AF142570.1 - 4 392
AF338733.1 - 4 376
AK026800.1 - 4 464
AK223303.1 - 4 492
AK225583.1 - 4 464
AY775789.1 - 3 273

View Full Product Details